Learn More
Sino Biological Recombinant Mouse TIM-1/HAVCR1 Protein (His Tag)
Supplier:  Sino Biological 50321M07H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse HAVCR1 (AAL35774.1) (Tyr 22-Thr 212) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1747