Learn More
Sino Biological Recombinant Mouse CD16/FCGR3/Fc gamma RIII Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 50326M08H100
Encompass_Preferred
A DNA sequence encoding the mouse FCGR3 (P08508-1) extracellular domain (Met 1-Thr 215) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1757