Learn More
Sino Biological Recombinant Mouse CD147/Basigin/EMMPRIN Protein (His Tag)
Supplier:  Sino Biological 50332M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse BSG isoform 2 (NP_001070652.1) (Met 1-Arg 209) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1771