Learn More
Aviva Systems Biology Corp SUGCT Antibody - C-terminal region (ARP62521_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62521_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 5033411D12Rik. It was validated on Western Blot. . Alternate names: D17907ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-702