Learn More
Sino Biological Recombinant Mouse TNF-alpha Protein (ECD)
Supplier:  Sino Biological 50349MNAE1
Encompass_Preferred
A DNA sequence encoding the soluble form of mouse TNF-α (NP_038721.1) (Leu 80-Leu 235) was expressed, with an initial Met at the N-terminus.
Catalog No. 50-162-1790