Learn More
Sino Biological Recombinant Mouse BAFFR/TNFRSF13C Protein (rFc Tag)
Supplier:  Sino Biological 50353M31H200
Encompass_Preferred
A DNA sequence encoding the mouse Tnfrsf13c (NP_082351.1) (Met1-Ala71) was expressed with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-162-1802