Learn More
Sino Biological Recombinant Mouse ANGPTL4 Protein (His Tag)
Supplier:  Sino Biological 50356M07H100
Encompass_Preferred
A DNA sequence encoding the mouse ANGPTL4 (Q9Z1P8) (Lys167-Ser410) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1806