Learn More
Sino Biological Recombinant Mouse ITK Protein (aa 351-619, His & GST Tag)
Supplier:  Sino Biological 50361M20B50
Encompass_Preferred
A DNA sequence encoding the mouse ITK / EMT (Q03526) protein kinase domain (Arg 351-Leu 619) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-1809