Learn More
Sino Biological Recombinant Mouse GLO1 Protein (His Tag)
Supplier:  Sino Biological 50364M07E100
Encompass_Preferred
A DNA sequence encoding the mouse GLO1 (NP_079650.3) (Ala 2-Ile 184) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-162-1812