Learn More
Sino Biological Recombinant Mouse Prolactin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50367M08B100
Encompass_Preferred
A DNA sequence encoding the mouse PRL (NP_035294.2) (Leu 32-Cys 228) was expressed,with an initial Met at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1816