Learn More
Sino Biological Recombinant Mouse MCP-1/CCL2 Protein (His Tag)
Supplier:  Sino Biological 50368M08B50
Encompass_Preferred
A DNA sequence encoding the mouse CCL2 (NP_035463.1) (Met1-Asn148) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1819