Learn More
Sino Biological Recombinant Mouse Beta-NGF Protein
Supplier:  Sino Biological 50385MNAC20
Encompass_Preferred
A DNA sequence encoding the mature form of mouse NGF (NP_001106168.1) (Ser 122-Gly 241) was expressed, with an initial Met at the C-terminus.
Catalog No. 50-162-1832