Learn More
Sino Biological Recombinant Mouse CD8 alpha Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50389M08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD8a (NP_001074579.1) (Met 1-Tyr 196) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1842