Learn More
Sino Biological Recombinant Mouse CD3 delta/CD3D Protein (hFc Tag)
Supplier:  Sino Biological 50390M02H100
Encompass_Preferred
A DNA sequence encoding the mouse CD3D (P04235) (Met1-Ala105) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1844