Learn More
Sino Biological Recombinant Mouse CD7 Protein (His Tag)
Supplier:  Sino Biological 50393M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CD7 (NP_033984.1) extracellular domain (Met 1-Pro 150) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1848