Learn More
Sino Biological Recombinant Mouse Progranulin Protein (His Tag)
Supplier:  Sino Biological 50396M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Progranulin (NP_032201.2) (Met 1-Leu 589) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1852