Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
SDP

Catalog No. 08801246 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
Diameter (Metric):
11.5 mm
29 mm
34 mm
Length (Metric):
1.1 mL/cm vol./length
5 m
6.4 mL/cm vol./length
Quantity:
5 m Tubing/Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric) Quantity
08-801-246 34 mm 5 m 5 m Tubing/Kit
08-700-163 29 mm 6.4 mL/cm vol./length 5 m Tubing/Kit
08-801-245 11.5 mm 1.1 mL/cm vol./length 5 m Tubing/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 08-801-246 Supplier Spectrum™ Supplier No. 132725T
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 1m or 5m of Spectra/Por 3 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)

Specifications

Product Type Trial Kit
MWCO 3.5 kDa
Membrane Regenerated Cellulose
Diameter (Metric) 34 mm
Diameter (English) 0.2 in.
Length (English) 16 ft.
Length (Metric) 5 m
Autoclavable Autoclavable
Capacity (Metric) 9.3 mL
Kit Contents 5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks
Quantity 5 m Tubing/Kit
For Use With (Application) ideal for small volume dialysis or evaluation of membrane for an application
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.