Learn More
Sino Biological Recombinant Mouse THSD1 Protein (His Tag)
Supplier:  Sino Biological 50400M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse THSD1 (NP_062522.1) (Met 1-Asn 412) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1856