Learn More
Sino Biological Recombinant Mouse Endoglin/CD105 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50407M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CD105 (NP_031958.2) extracellular domain (Met 1-Gly 580) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1862