Learn More
Sino Biological Recombinant Mouse CD31/PECAM-1 Protein (His Tag)
Supplier:  Sino Biological 50408M08H100
Encompass_Preferred
A DNA sequence encoding the mouse PECAM1 (Q08481-1) extracellular domain (Met 1-Lys 590) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1865