Learn More
Sino Biological Recombinant Mouse DR5/TRAIL R2 Protein (His & hFc Tag)
Supplier:  Sino Biological 50412M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TNFRSF10B (NP_064671.2) (Met 1-Ser 177) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1871