Learn More
Sino Biological Recombinant Mouse CD48 Protein (hFc Tag)
Supplier:  Sino Biological 50415M02H100
Encompass_Preferred
A DNA sequence encoding the mouse CD48 (BAE96323.1) (Met1-Arg216) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1877