Learn More
Sino Biological Recombinant Mouse CD48 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50415M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CD48 (NP_031675.1) without the propeptide, corresponding to the amino acids (Met 1-Arg 216) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1879