Learn More
Sino Biological Recombinant Mouse CD36 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50422M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD36 (NP_001153030.1) (Gly 30-Lys 439) was expressed, with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-162-1887