Learn More
Sino Biological Recombinant Mouse Serpin A11 Protein (His Tag)
Supplier:  Sino Biological 50435M08H100
Encompass_Preferred
A DNA sequence encoding the mouse SERPINA11 isoform 1 (NP_955018.2) (Met 1-Gly 424) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1899