Learn More
Hach Company Sension+ 5044 Reference Electrode for use with Ion Selective Electrodes ISE (ISE)
Catalog No. 501997964 Shop All Hach Company Products
Encompass
Sension+ 5044 is a double junction reference electrode with a glass body and internal gel electrolyte and 0.1M lithium acetate salt bridge. The reference probe has a large, replaceable ceramic diaphragm. The 5044 has a fixed 1 meter cable and banana connector for laboratory use with the Hach Sension+ MM374 Multimeter. The 5044 with lithium acetate electrolyte is recommended for use with all Sension+ half-cell ion selective electrodes (ISE) in drinking water, wastewater and general water quality applications.
Catalog No. 50-199-7964 Supplier Hach Company Supplier No. LZW5044.97.0002
Sension+ 5044 is a double junction reference electrode with a glass body and internal gel electrolyte and 0.1M lithium acetate salt bridge. The reference probe has a large, replaceable ceramic diaphragm. The 5044 has a fixed 1 meter cable and banana connector for laboratory use with the Hach Sension+ MM374 Multimeter. The 5044 with lithium acetate electrolyte is recommended for use with all Sension+ half-cell ion selective electrodes (ISE) in drinking water, wastewater and general water quality applications.