Learn More
Sino Biological Recombinant Mouse ICAM-1/CD54 Protein (His & hFc Tag)
Supplier:  Sino Biological 50440M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse ICAM1 (NP_034623.1) (Met 1-Asn 485) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1907