Learn More
Sino Biological Recombinant Mouse PDGF-A Protein (His Tag)
Supplier:  Sino Biological 50447M07Y50
Encompass_Preferred
A DNA sequence encoding the mouse PDGFA (NP_032834.1) (Ser87-Arg196) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1922