Learn More
Sino Biological Recombinant Mouse Periostin Protein (His Tag)
Supplier:  Sino Biological 50450M08H100
Encompass_Preferred
A DNA sequence encoding the full length of mouse POSTN (NP_056599.1) (Met 1-Gln 811) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1925