Learn More
Sino Biological Recombinant Mouse Prolactin Receptor Protein (His & hFc Tag)
Supplier:  Sino Biological 50457M03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse PRLR (NP_035299.4) (Met 1-Asp 229) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1929