Learn More
Sino Biological Recombinant Mouse Prolactin Receptor Protein (His Tag)
Supplier:  Sino Biological 50457M08H100
Encompass_Preferred
A DNA sequence encoding the mouse PRLR (P07321) (Met1-Asp229) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1931