Learn More
Sino Biological Recombinant Mouse PDGF-C Protein (hFc Tag)
Supplier:  Sino Biological 50458M01H100
Encompass_Preferred
A DNA sequence encoding the receptor-binding form of mouse PDGFC (Q8CI19) (Val 235-Gly 345) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-1933