Learn More
Sino Biological Recombinant Mouse PDGF-C Protein (His Tag)
Supplier:  Sino Biological 50458M07Y100
Encompass_Preferred
A DNA sequence encoding the mouse Pdgfc (NP_064355.1) (Val235-Gly345) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1936