Learn More
Sino Biological Recombinant Mouse SHP-2 Protein (His Tag)
Supplier:  Sino Biological 50462M08H200
Encompass_Preferred
A DNA sequence encoding the mouse PTPN11 (P35235-2) (Met1-Arg593) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1946