Learn More
Sino Biological Recombinant Mouse JAM-2/JAM-B Protein (hFc Tag)
Supplier:  Sino Biological 50464M02H100
Encompass_Preferred
A DNA sequence encoding the mouse JAM-2 (NP_076333.3) extracellular domain (Met 1-Asn 236) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1949