Learn More
Sino Biological Recombinant Mouse JAM-2/JAM-B Protein (His Tag)
Supplier:  Sino Biological 50464M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse JAM2 (NP_076333.3) (Met 1-Asn 236) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1951