Learn More
Sino Biological Recombinant Mouse ICOS Protein (rFc Tag)
Supplier:  Sino Biological 50466M31H100
Encompass_Preferred
A DNA sequence encoding the mouse Icos (NP_059508.2) (Met1-Leu144) was expressed with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-162-1957