Learn More
Sino Biological Recombinant Mouse Interferon alpha/beta receptor 1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50469M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse IFNAR1 (NP_034638.2) extracellular domain (Met 1-Thr 429) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1961