Learn More
Sino Biological Recombinant Mouse EGFL7 Protein (His Tag)
Supplier:  Sino Biological 50472M07B100
Encompass_Preferred
A DNA sequence encoding the mouse EGFL7 isoform 1 (Q9QXT5-1) (Glu 22-Leu 275) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-1963