Learn More
Sino Biological Recombinant Mouse Osteoactivin/GPNMB Protein (His Tag)
Supplier:  Sino Biological 50475M08H100
Encompass_Preferred
A DNA sequence encoding the mouse GPNMB (Q99P91) (Met1-Asn502) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-1970