Learn More
Sino Biological Recombinant Mouse CADM3 Protein (His Tag)
Supplier:  Sino Biological 50476M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CADM3 (NP_444429.1) extracellular domain (Met 1-His 328) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1972