Learn More
Sino Biological Recombinant Mouse EphB1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50479M08H200
Encompass_Preferred
A DNA sequence encoding the mouse EPHB1 (Q8CBF3-1) extracellular domain (Met 1-Leu 539) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1975