Learn More
Hach Company Sension+ 5048 portable multi-parameter electrode: pH, conductivity, ORP and temperature
Catalog No. 501997966 Shop All Hach Company Products
Encompass
Sension+ 5048 is a combination multi-parameter electrode for determination of pH, conductivity, ORP and temperature. The 5048 has a plastic body, non-refillable gel reference electrolyte and built-in temperature sensor. The electrode has a fixed 1 meter cable and MP8 connector dedicated for use with Hach Sension+ MM150 portable meters. The 5048 has a ceramic pin reference junction and glass (pH); platinum (conductivity & ORP); and Pt1000 (temperature) sensors. It is ideal for pH, conductivity, ORP and temperature measurements in general aqueous applications.
Catalog No. 50-199-7966 Supplier Hach Company Supplier No. LZW5048.97.0002
Sension+ 5048 is a combination multi-parameter electrode for determination of pH, conductivity, ORP and temperature. The 5048 has a plastic body, non-refillable gel reference electrolyte and built-in temperature sensor. The electrode has a fixed 1 meter cable and MP8 connector dedicated for use with Hach Sension+ MM150 portable meters. The 5048 has a ceramic pin reference junction and glass (pH); platinum (conductivity & ORP); and Pt1000 (temperature) sensors. It is ideal for pH, conductivity, ORP and temperature measurements in general aqueous applications.