Learn More
Sino Biological Recombinant Mouse EGF Protein (hFc Tag)
Supplier:  Sino Biological 50482M01H1
Encompass_Preferred
A DNA sequence encoding the mature form of mouse EGF (P01132) (Asn 977-Arg 1029) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-1978