Learn More
Sino Biological Recombinant Mouse EGF Protein
Supplier:  Sino Biological 50482MNAY100
Encompass_Preferred
A DNA sequence encoding the mouse EGF (P01132) (Asn977-Arg1029) was expressed and purified.
Catalog No. 50-162-1984