Learn More
Sino Biological Recombinant Mouse S100A4 Protein
Supplier:  Sino Biological 50484MNCH100
Encompass_Preferred
A DNA sequence encoding the mouse S100A4 (P07091)(Ala2-Lys101) was expressed and purified with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-162-1993