Learn More
Sino Biological Recombinant Mouse Clusterin Protein (His Tag)
Supplier:  Sino Biological 50485M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CLU (NP_038520.2) (Met 1-Glu 448) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-1995