Learn More
Sino Biological Recombinant Mouse SCF Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 50487M08B100
Encompass_Preferred
A DNA sequence encoding the amino acids (Met 1-Ala 189) of mouse KITL (P20826-1) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1999