Learn More
Sino Biological Recombinant Mouse RP105/CD180 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50491M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CD180 (NP_032559.2) extracellular domain (Met 1-Ser 626) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2009