Learn More
Sino Biological Recombinant Mouse SPARC Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50494M08H100
Encompass_Preferred
A DNA sequence encoding the mouse SPARC (NP_033268.1) (Met 1-Ile 302) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2015